• USA Home
  • 8.18404 - Methyl methoxyacetate

EMAIL THIS PAGE TO A FRIEND
8.18404 EMD Millipore

Methyl methoxyacetate

Methyl methoxyacetate for synthesis. CAS 6290-49-9, molar mass 104.11 g/mol., for synthesis

Synonym: Methoxyacetic acid methyl ester

  • CAS Number 6290-49-9

  • Linear Formula C₄H₈O₃

  • Molecular Weight 104.11 g/mol

  •  EC Number 228-539-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8184040100)
bp   129 - 131 °C (1013 hPa)
density   1.05 g/cm3 (20 °C)
flash point   35 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.049 - 1.051
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2918 99 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
RTECS 
AI8910000
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 3272 , 3, III
Declaration (transport by air) IATA-DGR 
UN 3272 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 3272 , 3, III
Categories of danger 
flammable
R Phrase 
R 10
Flammable.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?