EMAIL THIS PAGE TO A FRIEND
8.18409 EMD Millipore

(R)-(-)-Carvone

(R)-(-)-Carvone for synthesis. CAS 6485-40-1, molar mass 150.22 g/mol., for synthesis

Synonym: (R)-(-)-5-Isopropenyl-2-methyl-2-cyclohexenone, (R)-(-)-p-Mentha-6,8-dien-2-one, Carvol

  • CAS Number 6485-40-1

  • Linear Formula C₁₀H₁₄O

  • Molecular Weight 150.22 g/mol

  •  EC Number 229-352-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8184090100)
bp   231 °C (101.325 hPa)
solubility   29 - 79 g/l insoluble
density   0.96 g/cm3 (20 °C)
flash point   92 °C
LD 50 oral   LD50 Rat 1640 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.959 - 0.961
  Spec. rotation [α²0/D (undiluted): -63.0 - -60.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Hazard Statement(s) 
H317: May cause an allergic skin reaction.
Precautionary Statement(s) 
P280: Wear protective gloves.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
harmful
R Phrase 
R 43
May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?