• USA Home
  • 8.18415 - 3-Allyloxy-1,2-propanediol

EMAIL THIS PAGE TO A FRIEND
8.18415 EMD Millipore

3-Allyloxy-1,2-propanediol

3-Allyloxy-1,2-propanediol for synthesis. CAS No. 123-34-2, EC Number 204-620-4., for synthesis

Synonym: Glycerol 1-allyl ether

  • CAS Number 123-34-2

  • Linear Formula C₆H₁₂O₃

  • Molecular Weight 132.15 g/mol

  •  EC Number 204-620-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8184150100)
vapor pressure   0.004 hPa (20 °C)
density   1.07 g/cm3 (25 °C)
flash point   98 °C DIN 51758
pH   6.5 - 7.5 (1000 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 5400 mg/kg
LD 50 dermal   LD50 Rat 4300 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.068 - 1.070
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 49 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?