• USA Home
  • 8.18441 - 3'-Methoxyacetophenone

EMAIL THIS PAGE TO A FRIEND
8.18441 EMD Millipore

3'-Methoxyacetophenone

3'-Methoxyacetophenone for synthesis. CAS No. 586-37-8, EC Number 209-573-3., for synthesis

Synonym: 3-Acetylanisole

  • CAS Number 586-37-8

  • Linear Formula C₉H₁₀O₂

  • Molecular Weight 150.18 g/mol

  •  EC Number 209-573-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8184410050)
density   1.09 g/cm3 (20 °C)
flash point   110 °C
mp   -8 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.093 - 1.095
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 50 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?