EMAIL THIS PAGE TO A FRIEND
8.18445 EMD Millipore

Tryptamine

Tryptamine for synthesis. CAS 61-54-1, pH 9.7 (1 g/l, H₂O, 20 °C)., for synthesis

Synonym: 3-(2-Aminoethyl)indole, 2-(3-Indolyl)ethylamine

  • CAS Number 61-54-1

  • Linear Formula C₁₀H₁₂N₂

  • Molecular Weight 160.22 g/mol

  •  EC Number 200-510-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8184450010)
flash point   185 °C
autoignition temp.   491 °C
mp   116 °C
pH   11.07 (10 g/l, H₂O, 24.7 °C)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 98.0 %
  Melting range (lower value): ≥ 113 °C
  Melting range (upper value): ≤ 116 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Hazard Statement(s) 
H302: Harmful if swallowed.
H314: Causes severe skin burns and eye damage.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
NL 4020000
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 3259 , 8, II
Declaration (transport by air) IATA-DGR 
UN 3259 , 8, II
Declaration (transport by sea) IMDG-Code 
UN 3259 , 8, II, Segregation Group: 18 (Alkalis)
R Phrase 
R 22-34-37
Harmful if swallowed.Causes burns.Irritating to respiratory system.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?