EMAIL THIS PAGE TO A FRIEND
8.18451 EMD Millipore

Isoeugenol

Isoeugenol for synthesis. CAS 97-54-1, EC Number 202-590-7, chemical formula 2-(CH₃O)-4-(CH₃CH=CH)C₆H₃OH., for synthesis

Synonym: 4-Propenyl-2-methoxyphenol, 2-Methoxy-4-propenylphenol

  • CAS Number 97-54-1

  • Linear Formula C₁₀H₁₂O₂

  • Molecular Weight 164.21 g/mol

  •  EC Number 202-590-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8184510100)
bp   266 - 268 °C (1013 hPa)
density   1.09 g/cm3 (20 °C)
flash point   150 °C
mp   19 - 23 °C
LD 50 oral   LD50 Rat 1560 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay of cis-trans-Isomers (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.085 - 1.089
  trans-Isoeugenol (GC, area%): ≥ 85.0 % (a/a)
  cis-Isoeugenol (GC, area%): ≤ 15.0 % (a/a)
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 50 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24/25
Avoid contact with skin and eyes.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?