EMAIL THIS PAGE TO A FRIEND
8.18453 EMD Millipore

(-)-Cinchonidine

(-)-Cinchonidine for resolution of racemates for synthesis. CAS 485-71-2, pH 9 (0.2 g/l, H₂O, 20 °C)., for resolution of racemates for synthesis

  • CAS Number 485-71-2

  • Linear Formula C₁₉H₂₂N₂O

  • Molecular Weight 294.39 g/mol

  •  EC Number 207-622-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8184530025)
solubility   0.25 g/l
mp   199 - 202 °C
pH   9 (0.2 g/l, H₂O, 20 °C)
bulk density   155 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 99.0 %
  Melting range (lower value): ≥ 200 °C
  Melting range (upper value): ≤ 204 °C
  Spec. rotation [α²0/D (c=5 in ethanol): -108.0 - -104.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2939 20 00
Precautionary Statement(s) 
P260: Do not breathe dust.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
S Phrase 
S 22
Do not breathe dust.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?