EMAIL THIS PAGE TO A FRIEND
8.18456 EMD Millipore

4-Methylanisole

4-Methylanisole for synthesis. CAS 104-93-8, chemical formula 4-(CH₃)C₆H₄OCH₃., for synthesis

Synonym: p-Cresol methyl ether

  • CAS Number 104-93-8

  • Linear Formula C₈H₁₀O

  • Molecular Weight 122.17 g/mol

  •  EC Number 203-253-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 ml in Glass bottle (8184560250)
bp   174 - 177 °C (1013 hPa)
expl. lim.     1.1 - 8.3 %(V)
vapor pressure   2 hPa (20 °C)
density   0.97 g/cm3 (20 °C)
flash point   58 °C
autoignition temp.   485 °C
mp   -50 °C
LD 50 oral   LD50 Rat 1920 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.968 - 0.970
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
H315: Causes skin irritation.
Precautionary Statement(s) 
P210: Keep away from heat.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
harmful, irritant
R Phrase 
R 22-38
Harmful if swallowed.Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?