• USA Home
  • 8.18457 - L-(+)-α-Phenylglycine

EMAIL THIS PAGE TO A FRIEND
8.18457 EMD Millipore

L-(+)-α-Phenylglycine

L-(+)-α-Phenylglycine (for resolution of racemates) for synthesis. CAS 2935-35-5, molar mass 151.16 g/mol., (for resolution of racemates) for synthesis

  • CAS Number 2935-35-5

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 220-909-8

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8184570025)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HClO₄): ≥ 99.0 %
  Spec. rotation [α²0/D (c=1 in HCl 1mol/l): +155.0 - +157.0 °
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?