EMAIL THIS PAGE TO A FRIEND
8.18458 EMD Millipore

Triphenylsilane

Triphenylsilane for synthesis. CAS 789-25-3, EC Number 212-333-0, chemical formula (C₆H₅)₃SiH., for synthesis

  • CAS Number 789-25-3

  • Linear Formula C₁₈H₁₆Si

  • Molecular Weight 260.41 g/mol

  •  EC Number 212-333-0

Purchase

Properties

packaging   25 g in Glass bottle (8184580025)
bp   151 - 153 °C (3 hPa)
flash point   76 °C
mp   43 - 45 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 43 °C
  Melting range (upper value): ≤ 45 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2931 90 00
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?