• USA Home
  • 8.18460 - 1,2-Bis(cyanomethyl)-benzene

EMAIL THIS PAGE TO A FRIEND
8.18460 EMD Millipore

1,2-Bis(cyanomethyl)-benzene

1,2-Bis(cyanomethyl)-benzene for synthesis. CAS 613-73-0, chemical formula C₆H₄-1,2-(CH₂CN)₂., for synthesis

Synonym: o-Xylylene dicyanide, 1,2-Phenylenediacetonitrile

  • CAS Number 613-73-0

  • Linear Formula C₁₀H₈N₂

  • Molecular Weight 156.18 g/mol

  •  EC Number 210-351-3

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8184600010)
mp   57 - 59 °C
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 57 °C
  Melting range (upper value): ≤ 59 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 1 slightly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, III
Categories of danger 
toxic
R Phrase 
R 23/24/25
Toxic by inhalation, in contact with skin and if swallowed.
S Phrase 
S 26-36/37-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?