• USA Home
  • 8.18468 - 1-Acetylnaphthalene

EMAIL THIS PAGE TO A FRIEND
8.18468 EMD Millipore

1-Acetylnaphthalene

1-Acetylnaphthalene for synthesis. CAS 941-98-0, chemical formula C₁₀H₇C(O)CH₃., for synthesis

Synonym: Methyl 1-naphthyl ketone

  • CAS Number 941-98-0

  • Linear Formula C₁₂H₁₀O

  • Molecular Weight 170.21 g/mol

  •  EC Number 213-384-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8184680050)
bp   302 °C (1013 hPa)
vapor pressure   <0.1 hPa (20 °C)
solubility   0.2 g/l
density   1.12 g/cm3 (20 °C)
flash point   >100 °C
mp   9 - 11 °C
pH   6.5 - 7.0 (H₂O)
LD 50 oral   LD50 Rat 1560 mg/kg
LD 50 dermal   LD50 Rabbit > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 96.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.117 - 1.119
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 39 00
Hazard Statement(s) 
H302: Harmful if swallowed.
H412: Harmful to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 22-52/53
Harmful if swallowed.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 61
Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?