EMAIL THIS PAGE TO A FRIEND
8.18472 EMD Millipore

4-Isopropylaniline

4-Isopropylaniline for synthesis. CAS 99-88-7, chemical formula 4-(NH₂)-C₆H₄-CH(CH₃)2., for synthesis

Synonym: Cumidine, p-Aminocumene

  • CAS Number 99-88-7

  • Linear Formula C₉H₁₃N

  • Molecular Weight 135.21 g/mol

  •  EC Number 202-797-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8184720050)
bp   226 - 227 °C (1013 hPa)
vapor pressure   0.1 hPa (20 °C)
solubility   2.2 g/l
density   0.98 g/cm3 (20 °C)
flash point   92 °C
autoignition temp.   540 °C
mp   -63 °C
pH   7 (2.2 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 757 mg/kg
LD 50 dermal   LD50 Rat 1030 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.950 - 0.951
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 49 00
Hazard Statement(s) 
H302 + H312: Harmful if swallowed or in contact with skin
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
8A Combustible, corrosive hazardous materials
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 1760 , 8, III
Declaration (transport by air) IATA-DGR 
UN 1760 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 1760 , 8, III
Categories of danger 
harmful, corrosive
R Phrase 
R 21/22-34
Harmful in contact with skin and if swallowed.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?