• USA Home
  • 8.18476 - Monomethyl 5-nitroisophthalate

EMAIL THIS PAGE TO A FRIEND
8.18476 EMD Millipore

Monomethyl 5-nitroisophthalate

Monomethyl 5-nitroisophthalate for synthesis. CAS 1955-46-0, EC Number 217-793-6, chemical formula 5-(NO₂)-3-(COOH)C₆H₃COOCH₃., for synthesis

Synonym: 5-Nitroisophthalic acid monomethyl ester

  • CAS Number 1955-46-0

  • Linear Formula C₉H₇NO₆

  • Molecular Weight 225.16 g/mol

  •  EC Number 217-793-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8184760050)
mp   177 - 180 °C
bulk density   750 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 177 °C
  Melting range (upper value): ≤ 180 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2917 39 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?