• USA Home
  • 8.18480 - 6-Amino-1,4-benzodioxane

EMAIL THIS PAGE TO A FRIEND
8.18480 EMD Millipore

6-Amino-1,4-benzodioxane

6-Amino-1,4-benzodioxane for synthesis. CAS 22013-33-8, molar mass 151.16 g/mol., for synthesis

Synonym: 3.4-Ethylenedioxyaniline

  • CAS Number 22013-33-8

  • Linear Formula C₈H₉NO₂

  • Molecular Weight 151.16 g/mol

  •  EC Number 244-718-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8184800010)
bp   156 - 158 °C (12 hPa)
density   1.25 g/cm3 (20 °C)
flash point   >112 °C
mp   30 °C
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.251 - 1.253
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Hazard Statement(s) 
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 2810 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2810 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2810 , 6.1, II
Categories of danger 
irritant
R Phrase 
R 38
Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?