• USA Home
  • 8.18507 - 4-Amino-1,3-dimethyluracil

EMAIL THIS PAGE TO A FRIEND
8.18507 EMD Millipore

4-Amino-1,3-dimethyluracil

4-Amino-1,3-dimethyluracil for synthesis. CAS 6642-31-5, pH 6.9 (100 g/l, H₂O, 20 °C)., for synthesis

Synonym: 1,3-Dimethyl-4-aminouracil, 1,3-Dimethyl-2,6-dioxo-4-aminotetrahydropyrimidine

  • CAS Number 6642-31-5

  • Linear Formula C₆H₉N₃O₂

  • Molecular Weight 155.15 g/mol

  •  EC Number 229-662-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 g in Plastic bottle (8185070050)
solubility   6 g/l
mp   294 - 296 °C
pH   6.9 (100 g/l, H₂O, 20 °C)
bulk density   400 kg/m3
LD 50 oral   LD50 Rat > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 59 95
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?