• USA Home
  • 8.18510 - Diethyl aminomalonate hydrochloride

EMAIL THIS PAGE TO A FRIEND
8.18510 EMD Millipore

Diethyl aminomalonate hydrochloride

Diethyl aminomalonate hydrochloride for synthesis. CAS 13433-00-6, chemical formula (H₅C₂OOC)₂CHNH₂ * HCl., for synthesis

Synonym: Aminomalonic acid diethyl ester hydrochloride

  • CAS Number 13433-00-6

  • Linear Formula C₇H₁₄ClNO₄

  • Molecular Weight 211.64 g/mol

  •  EC Number 236-556-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8185100025)
mp   165 - 170 °C
bulk density   400 kg/m3
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 99.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 49 85
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?