• USA Home
  • 8.18519 - 1,2-Methylenedioxybenzene

EMAIL THIS PAGE TO A FRIEND
8.18519 EMD Millipore

1,2-Methylenedioxybenzene

1,2-Methylenedioxybenzene for synthesis. CAS 274-09-9, molar mass 122.12 g/mol., for synthesis

Synonym: 1,3-Benzodioxol

  • CAS Number 274-09-9

  • Linear Formula C₇H₆O₂

  • Molecular Weight 122.12 g/mol

  •  EC Number 205-992-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8185190050)
bp   172 - 175 °C (1013 hPa)
vapor pressure   16 hPa (25 °C)
solubility   2 g/l
density   1.18 g/cm3 (20 °C)
flash point   61 °C
mp   -18 °C
LD 50 oral   LD50 Rat 580 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.184 - 1.186
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2932 99 00
Hazard Statement(s) 
H302: Harmful if swallowed.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Categories of danger 
harmful
R Phrase 
R 22
Harmful if swallowed.
S Phrase 
S 24
Avoid contact with skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?