• USA Home
  • 8.18558 - (4-Methoxyphenyl) acetone

EMAIL THIS PAGE TO A FRIEND
8.18558 EMD Millipore

(4-Methoxyphenyl) acetone

(4-Methoxyphenyl) acetone for synthesis. CAS No. 122-84-9, EC Number 204-578-7., for synthesis

Synonym: 1-(4-Methoxyphenyl)-2-propanone, 4-Methoxybenzyl methyl ketone

  • CAS Number 122-84-9

  • Linear Formula C₁₀H₁₂O₂

  • Molecular Weight 164.21 g/mol

  •  EC Number 204-578-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8185580050)
bp   134 - 136 °C (13 hPa)
density   1.07 g/cm3 (20 °C)
flash point   135 °C
LD 50 oral   LD50 Rat 3330 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.5 % (a/a)
  Density (d 20 °C/ 4 °C): 1.064 - 1.067
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 50 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?