• USA Home
  • 8.18564 - 4-Phenylcyclohexanone

EMAIL THIS PAGE TO A FRIEND
8.18564 EMD Millipore

4-Phenylcyclohexanone

4-Phenylcyclohexanone for synthesis. CAS 4894-75-1, molar mass 174.24 g/mol., for synthesis

  • CAS Number 4894-75-1

  • Linear Formula C₁₂H₁₄O

  • Molecular Weight 174.24 g/mol

  •  EC Number 225-517-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8185640025)
bp   158 - 160 °C (16 hPa)
density   0.97 g/cm3 (25 °C)
flash point   >100 °C
mp   76 - 78 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 76 °C
  Melting range (upper value): ≤ 78 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 39 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?