• USA Home
  • 8.18566 - 1,4-Cyclohexanedione

EMAIL THIS PAGE TO A FRIEND
8.18566 EMD Millipore

1,4-Cyclohexanedione

1,4-Cyclohexanedione for synthesis. CAS 637-88-7, molar mass 112.12 g/mol., for synthesis

  • CAS Number 637-88-7

  • Linear Formula C₆H₈O₂

  • Molecular Weight 112.12 g/mol

  •  EC Number 211-306-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8185660025)
bp   111 - 113 °C (26 hPa)
mp   76 - 78 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 76 °C
  Melting range (upper value): ≤ 78 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?