EMAIL THIS PAGE TO A FRIEND
8.18570 EMD Millipore

Farnesol

Farnesol for synthesis. CAS 4602-84-0, EC Number 225-004-1, chemical formula (CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(CH₃)=CHCH₂OH., for synthesis

Synonym: 3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol

  • CAS Number 4602-84-0

  • Linear Formula C₁₅H₂₆O

  • Molecular Weight 222.37 g/mol

  •  EC Number 225-004-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8185700025)
density   0.89 g/cm3 (20 °C)
flash point   96 °C
autoignition temp.   240 °C
LD 50 oral   LD50 Rat 6000 mg/kg
storage temp.   Store below +30°C.
specifications   Assay isomers (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.886 - 0.888
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 22 00
Hazard Statement(s) 
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RTECS 
JR4979000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
UN 3082 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3082 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3082 , 9, III, Marine Pollutant: P
R Phrase 
R 36/38-43
Irritating to eyes and skin.May cause sensitisation by skin contact.
S Phrase 
S 24-37
Avoid contact with skin.Wear suitable gloves.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?