EMAIL THIS PAGE TO A FRIEND
8.18575 EMD Millipore

2-Undecanol

2-Undecanol for synthesis. CAS No. 1653-30-1, EC Number 216-722-6., for synthesis

Synonym: 2-Hendecanol

  • CAS Number 1653-30-1

  • Linear Formula C₁₁H₂₄O

  • Molecular Weight 172.31 g/mol

  •  EC Number 216-722-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8185750050)
bp   228 - 229 °C (1013 hPa)
density   0.83 g/cm3 (20 °C)
flash point   107 °C
mp   1 - 3 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.826 - 0.828
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2905 19 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?