EMAIL THIS PAGE TO A FRIEND
8.18576 EMD Millipore

3-Nonanone

3-Nonanone for synthesis. CAS No. 925-78-0, EC Number 213-125-2., for synthesis

Synonym: Ethyl hexyl ketone

  • CAS Number 925-78-0

  • Linear Formula C₉H₁₈O

  • Molecular Weight 142.24 g/mol

  •  EC Number 213-125-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   50 ml in Glass bottle (8185760050)
bp   185 - 186 °C (1013 hPa)
density   0.82 g/cm3 (20 °C)
flash point   63 °C
mp   -8 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.824 - 0.827
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 19 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?