EMAIL THIS PAGE TO A FRIEND
8.18585 EMD Millipore

2-Allylphenol

2-Allylphenol for synthesis. CAS 1745-81-9, pH 6 - 7 (100 g/l, H₂O, 20 °C)., for synthesis

  • CAS Number 1745-81-9

  • Linear Formula C₉H₁₀O

  • Molecular Weight 134.18 g/mol

  •  EC Number 217-119-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   215 - 223 °C (1013 hPa)
vapor pressure   1.6 hPa (50 °C)
solubility   7 g/l
density   1.02 g/cm3 (20 °C)
flash point   99 °C
mp   -2 °C
pH   6 - 7 (100 g/l, H₂O, 20 °C)
LD 50 oral   LD50 Rat 205 mg/kg
LD 50 dermal   LD50 Rat 770 - 1026 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.020 - 1.022
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 19 90
Hazard Statement(s) 
H301 + H311: Toxic if swallowed or in contact with skin
H314: Causes severe skin burns and eye damage.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
SJ3850000
Storage class 
6.1 C Combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Corrosive  Corrosive
Declaration (railroad and road) ADR, RID 
UN 2922 , 8 (6.1), III
Declaration (transport by air) IATA-DGR 
UN 2922 , 8 (6.1), III
Declaration (transport by sea) IMDG-Code 
UN 2922 , 8 (6.1), III
Categories of danger 
harmful, corrosive
R Phrase 
R 21/22-34
Harmful in contact with skin and if swallowed.Causes burns.
S Phrase 
S 26-36/37/39-45
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?