• USA Home
  • 8.18589 - 4-Nitrobenzonitrile

EMAIL THIS PAGE TO A FRIEND
8.18589 EMD Millipore

4-Nitrobenzonitrile

4-Nitrobenzonitrile for synthesis. CAS 619-72-7, chemical formula 4(NO₂)C₆H₄CN., for synthesis

  • CAS Number 619-72-7

  • Linear Formula C₇H₄N₂O₂

  • Molecular Weight 148.12 g/mol

  •  EC Number 210-610-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8185890025)
vapor pressure   0.0029 hPa (25 °C)
solubility   1.65 g/l slightly soluble
mp   145 - 147 °C
LD 50 oral   LD50 Rat 30 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 145 °C
  Melting range (upper value): ≤ 147 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2926 90 70
Hazard Statement(s) 
H300: Fatal if swallowed.
Precautionary Statement(s) 
P260: Do not breathe dust.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK 
WGK 2 water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Declaration (railroad and road) ADR, RID 
UN 3439 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 3439 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 3439 , 6.1, II
Categories of danger 
toxic
R Phrase 
R 25
Toxic if swallowed.
S Phrase 
S 22-36/37-45
Do not breathe dust.Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?