• USA Home
  • 8.18593 - 5-Fluoro-2-methylaniline

EMAIL THIS PAGE TO A FRIEND
8.18593 EMD Millipore

5-Fluoro-2-methylaniline

5-Fluoro-2-methylaniline for synthesis. CAS No. 367-29-3, EC Number 206-689-6., for synthesis

Synonym: 2-Amino-4-fluorotoluene, 2-Methyl-5-fluoroaniline

  • CAS Number 367-29-3

  • Linear Formula C₇H₈FN

  • Molecular Weight 125.15 g/mol

  •  EC Number 206-689-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Plastic bottle (8185930010)
bp   94 - 96 °C (20 hPa)
density   1.13 g/cm3 (20 °C)
flash point   100 °C
mp   36 - 38 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 35 °C
  Melting range (upper value): ≤ 38 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2921 43 00
Storage class 
4.1B Flammable solid hazardous materials
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 1325 , 4.1, II
Declaration (transport by air) IATA-DGR 
UN 1325 , 4.1, II
Declaration (transport by sea) IMDG-Code 
UN 1325 , 4.1, II
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?