• USA Home
  • 8.18594 - 2-Chlorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.18594 EMD Millipore

2-Chlorobenzyl alcohol

2-Chlorobenzyl alcohol for synthesis. CAS 17849-38-6, chemical formula 2-(Cl)C₆H₄CH₂OH., for synthesis

  • CAS Number 17849-38-6

  • Linear Formula C₇H₇ClO

  • Molecular Weight 142.58 g/mol

  •  EC Number 241-801-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8185940025)
bp   229 - 231 °C
mp   68 - 71 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 68 °C
  Melting range (upper value): ≤ 71 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2906 29 00
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?