• USA Home
  • 8.18596 - 2-Methylbenzimidazole

EMAIL THIS PAGE TO A FRIEND
8.18596 EMD Millipore

2-Methylbenzimidazole

2-Methylbenzimidazole for synthesis. CAS 615-15-6, molar mass 132.17 g/mol., for synthesis

  • CAS Number 615-15-6

  • Linear Formula C₈H₈N₂

  • Molecular Weight 132.17 g/mol

  •  EC Number 210-411-9

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Plastic bottle (8185960100)
mp   174 - 177 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 174 °C
  Melting range (upper value): ≤ 177 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?