• USA Home
  • 8.18616 - 4,4'-Dimethoxytriphenylmethyl chloride

EMAIL THIS PAGE TO A FRIEND
8.18616 EMD Millipore

4,4'-Dimethoxytriphenylmethyl chloride

4,4'-Dimethoxytriphenylmethyl chloride for synthesis. CAS 40615-36-9, chemical formula C₆H₅(CH₃OC₆H₄)₂CCl., for synthesis

Synonym: 4,4'-Dimethoxytrityl chloride

  • CAS Number 40615-36-9

  • Linear Formula C₂₁H₁₉ClO₂

  • Molecular Weight 338.83 g/mol

  •  EC Number 255-002-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8186160005)
mp   120 - 123 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 97.0 % (a/a)
  Melting range (lower value): ≥ 121 °C
  Melting range (upper value): ≤ 125 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?