• USA Home
  • 8.18620 - 2-tert-Butyl-4-methylphenol

EMAIL THIS PAGE TO A FRIEND
8.18620 EMD Millipore

2-tert-Butyl-4-methylphenol

2-tert-Butyl-4-methylphenol for synthesis. CAS No. 2409-55-4, EC Number 219-314-6., for synthesis

Synonym: 2-tert-Butyl-p-cresol

  • CAS Number 2409-55-4

  • Linear Formula C₁₁H₁₆O

  • Molecular Weight 164.24 g/mol

  •  EC Number 219-314-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   500 g in Plastic bottle (8186200500)
bp   237 °C (1013 hPa)
vapor pressure   0.02 hPa (20 °C)
density   0.92 g/cm3 (75 °C)
flash point   105 °C
mp   49 - 52 °C
pH   4 - 5 (H₂O, 20 °C) (saturated solution)
LD 50 oral   LD50 Rat 2390 mg/kg
LD 50 dermal   LD50 Rabbit 2200 mg/kg
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 49 °C
  Melting range (upper value): ≤ 53 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 19 90
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 36/38
Irritating to eyes and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?