• USA Home
  • 8.18622 - (2-Chloro-1,1-dimethylethyl)benzene

EMAIL THIS PAGE TO A FRIEND
8.18622 EMD Millipore

(2-Chloro-1,1-dimethylethyl)benzene

(2-Chloro-1,1-dimethylethyl)benzene for synthesis. CAS 515-40-2, chemical formula C₆H₅C(CH₃)₂CH₂Cl., for synthesis

Synonym: 1-Chloro-2-methyl-2-phenylpropane, 2-(Chloromethyl)-2-phenylpropane, Neophyl chloride

  • CAS Number 515-40-2

  • Linear Formula C₁₀H₁₃Cl

  • Molecular Weight 168.67 g/mol

  •  EC Number 208-197-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   222 °C (1013 hPa)
vapor pressure   0.08 hPa (20 °C)
density   1.04 g/cm3 (20 °C)
flash point   92 °C
autoignition temp.   458 °C
mp   <-60 °C
LD 50 oral   LD50 Rat 3228 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.041 - 1.043
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H315: Causes skin irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Categories of danger 
irritant
R Phrase 
R 38
Irritating to skin.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?