• USA Home
  • 8.18623 - 2,6-Di-tert-butylphenol

EMAIL THIS PAGE TO A FRIEND
8.18623 EMD Millipore

2,6-Di-tert-butylphenol

2,6-Di-tert-butylphenol for synthesis. CAS 128-39-2, chemical formula 2,6-(CH₃)₃C₂C₆H₃OH., for synthesis

  • CAS Number 128-39-2

  • Linear Formula C₁₄H₂₂O

  • Molecular Weight 206.32 g/mol

  •  EC Number 204-884-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8186230100)
bp   253 °C (1013 hPa)
vapor pressure   <0.01 hPa (20 °C)
solubility   0.003 g/l
density   0.914 g/cm3 (20 °C)
flash point   120 °C
mp   34 - 37 °C
LD 50 oral   LD50 Rat > 5000 mg/kg
LD 50 dermal   LD50 Rabbit > 10000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 34 °C
  Melting range (upper value): ≤ 37 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 19 90
Hazard Statement(s) 
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P262: Do not get in eyes, on skin, or on clothing.
P273: Avoid release to the environment.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3077 , 9, III
Declaration (transport by air) IATA-DGR 
UN 3077 , 9, III
Declaration (transport by sea) IMDG-Code 
UN 3077 , 9, III, Marine Pollutant: P
Categories of danger 
dangerous for the environment
R Phrase 
R 51/53
Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24/25-61
Avoid contact with skin and eyes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?