• USA Home
  • 8.18625 - 2-Phenylethyl acetate

EMAIL THIS PAGE TO A FRIEND
8.18625 EMD Millipore

2-Phenylethyl acetate

2-Phenylethyl acetate for synthesis. CAS No. 103-45-7, EC Number 203-113-5., for synthesis

Synonym: Acetic acid 2-phenylethyl ester

  • CAS Number 103-45-7

  • Linear Formula C₁₀H₁₂O₂

  • Molecular Weight 164.2 g/mol

  •  EC Number 203-113-5

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   224 °C (1013 hPa)
density   0.984 g/cm3 (20 °C)
flash point   107 °C
autoignition temp.   480 °C
mp   -31 °C
LD 50 oral   LD50 Rat 3670 mg/kg
LD 50 dermal   LD50 Rabbit 6210 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.032 - 1.034
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 39 00
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?