• USA Home
  • 8.18626 - 6-Methyl-5-hepten-2-one

EMAIL THIS PAGE TO A FRIEND
8.18626 EMD Millipore

6-Methyl-5-hepten-2-one

6-Methyl-5-hepten-2-one for synthesis. CAS No. 110-93-0, EC Number 203-816-7., for synthesis

Synonym: Methyl isohexenyl ketone

  • CAS Number 110-93-0

  • Linear Formula C₈H₁₄O

  • Molecular Weight 126.2 g/mol

  •  EC Number 203-816-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   172 - 174 °C
expl. lim.     1.1 - 7.3 %(V)
vapor pressure   1 hPa (20 °C)
solubility   3.02 g/l
density   0.85 g/cm3 (20 °C)
flash point   56 °C
autoignition temp.   250 °C DIN 51794
mp   -67 °C
pH   6.6 (3 g/l, H₂O, 25 °C)
LD 50 oral   LD50 Rat 3500 mg/kg
LD 50 dermal   LD50 Rat > 5000 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.850 - 0.860
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2914 19 90
Hazard Statement(s) 
H226: Flammable liquid and vapour.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS02  GHS02
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?