• USA Home
  • 8.18762 - 3-Aminobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
8.18762 EMD Millipore

3-Aminobenzyl alcohol

3-Aminobenzyl alcohol for synthesis. CAS No. 1877-77-6, EC Number 217-514-8., for synthesis

Synonym: 3-(Hydroxymethyl)aniline

  • CAS Number 1877-77-6

  • Linear Formula C₇H₉NO

  • Molecular Weight 123.16 g/mol

  •  EC Number 217-514-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8187620025)
mp   91 - 94 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (HPLC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 91 °C
  Melting range (upper value): ≤ 94 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2922 19 00
RTECS 
DN3154450
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?