EMAIL THIS PAGE TO A FRIEND
8.18768 EMD Millipore

1,3-Cyclooctadiene

1,3-Cyclooctadiene for synthesis. CAS 1700-10-3, molar mass 108.18 g/mol., for synthesis

  • CAS Number 1700-10-3

  • Linear Formula C₈H₁₂

  • Molecular Weight 108.18 g/mol

  •  EC Number 216-929-1

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   142 - 144 °C (1013 hPa)
vapor pressure   22 hPa (40 °C)
density   0.87 g/cm3 (20 °C)
flash point   25 °C
autoignition temp.   400 °C
mp   -53 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 97.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.870 - 0.872
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2902 19 00
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H304: May be fatal if swallowed and enters airways.
Precautionary Statement(s) 
P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking.
P301 + P310: IF SWALLOWED: Immediately call a POISON CENTER/doctor.
P331: Do NOT induce vomiting.
Storage class 
3 Flammable liquids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS08  GHS08
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2520 , 3, III
Declaration (transport by air) IATA-DGR 
UN 2520 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 2520 , 3, III
Categories of danger 
flammable, harmful
R Phrase 
R 10-65
Flammable.Harmful: may cause lung damage if swallowed.
S Phrase 
S 62
If swallowed, do not induce vomiting: seek medical advice immediately and show this container or label.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?