EMAIL THIS PAGE TO A FRIEND
8.20123 EMD Millipore

Benzanilide

Benzanilide for synthesis. CAS No. 93-98-1, EC Number 202-292-7., for synthesis

Synonym: N-Benzoylaniline

  • CAS Number 93-98-1

  • Linear Formula C₁₃H₁₁NO

  • Molecular Weight 197.23 g/mol

  •  EC Number 202-292-7

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 g in Glass bottle (8201230005)
mp   162 - 165 °C
bulk density   400 kg/m3
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 162 °C
  Melting range (upper value): ≤ 165 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2924 29 70
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?