• USA Home
  • 8.20130 - Benzohydroxamic acid

EMAIL THIS PAGE TO A FRIEND
8.20130 EMD Millipore

Benzohydroxamic acid

Benzohydroxamic acid for synthesis. CAS No. 495-18-1, EC Number 207-797-6., for synthesis

Synonym: N-Hydroxybenzamide, Phenylhydroxamic acid

  • CAS Number 495-18-1

  • Linear Formula C₇H₇NO₂

  • Molecular Weight 137.13 g/mol

  •  EC Number 207-797-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8201300025)
solubility   2.2 g/l
mp   125 - 127 °C
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 125 °C
  Melting range (upper value): ≤ 128 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?