EMAIL THIS PAGE TO A FRIEND
8.20148 EMD Millipore

Benzyl thiocyanate

Benzyl thiocyanate for synthesis. CAS No. 3012-37-1, EC Number 221-144-2., for synthesis

Synonym: Thiocyanic acid benzyl ester, Benzyl rhodanide

  • CAS Number 3012-37-1

  • Linear Formula C₈H₇NS

  • Molecular Weight 149.21 g/mol

  •  EC Number 221-144-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   250 g in Glass bottle (8201480250)
solubility   0.1 g/l
density   1.32 g/cm3 (20 °C)
flash point   110 °C
mp   38 - 41 °C
shipped in   ambient
storage temp.   Store at <= 20°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 38 °C
  Melting range (upper value): ≤ 41 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2930 90 98
Hazard Statement(s) 
H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
H412: Harmful to aquatic life with long lasting effects.
EUH032: Contact with acids liberates very toxic gas.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
RTECS 
XK8155000
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
6
Sulfides and sulfites as well as other reducing substances, inorganic cyanides and isonitriles: Stir into sodium hypochlorite solution (Cat. No. 105614) and, if necessary, leave to react for several days. Caution: some substances may show a violent reaction! Draw off any toxic or flammable gases that may be formed. Excess oxidant should be neutralized with sodium thiosulfate (Cat. No. 106513). Container D or E.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 2811 , 6.1, II
Declaration (transport by air) IATA-DGR 
UN 2811 , 6.1, II
Declaration (transport by sea) IMDG-Code 
UN 2811 , 6.1, II
Categories of danger 
harmful, dangerous for the environment
R Phrase 
R 20/21/22-32-52/53
Harmful by inhalation, in contact with skin and if swallowed.Contact with acids liberates very toxic gas.Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 13-61
Keep away from food, drink and animal feedingstuffs.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?