• USA Home
  • 8.20189 - Ethyl 4-bromobutyrate

EMAIL THIS PAGE TO A FRIEND
8.20189 EMD Millipore

Ethyl 4-bromobutyrate

Ethyl 4-bromobutyrate for synthesis. CAS 2969-81-5, EC Number 221-005-6, chemical formula Br(CH₂)₃COOC₂H₅., for synthesis

Synonym: 4-Bromobutyric acid ethyl ester

  • CAS Number 2969-81-5

  • Linear Formula C₆H₁₁BrO₂

  • Molecular Weight 195.05 g/mol

  •  EC Number 221-005-6

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 ml in Glass bottle (8201890025)
bp   80 - 82 °C (13 hPa)
density   1.35 g/cm3 (20 °C)
flash point   100 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 95.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.350 - 1.355
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2915 90 70
Hazard Statement(s) 
H315: Causes skin irritation.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
Precautionary Statement(s) 
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Irritant  Irritant
Declaration (railroad and road) ADR, RID 
Kein Gefahrgut
Declaration (transport by air) IATA-DGR 
UN 3334 , 9, III
Declaration (transport by sea) IMDG-Code 
No Dangerous Good
Categories of danger 
irritant
R Phrase 
R 36/37/38
Irritating to eyes, respiratory system and skin.
S Phrase 
S 26
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?