• USA Home
  • 8.20197 - 1-Bromo-3-fluorobenzene

EMAIL THIS PAGE TO A FRIEND
8.20197 EMD Millipore

1-Bromo-3-fluorobenzene

1-Bromo-3-fluorobenzene for synthesis. CAS 1073-06-9, chemical formula 1-(Br)-3-(F)C₆H₄., for synthesis

  • CAS Number 1073-06-9

  • Linear Formula C₆H₄BrF

  • Molecular Weight 175 g/mol

  •  EC Number 214-023-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 ml in Glass bottle (8201970010)
bp   148 - 150 °C (1013 hPa)
solubility   0.4 g/l
density   1.59 g/cm3 (20 °C)
flash point   38 °C
mp   -8 °C
LD 50 oral   LD50 Rat 670 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.591 - 1.595
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H332: Harmful if inhaled.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P210: Keep away from heat.
P273: Avoid release to the environment.
P280: Wear protective gloves.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Storage class 
3 Flammable liquids
WGK 
WGK 2 water endangering
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable, harmful, irritant, sensitizing, dangerous for the environment
R Phrase 
R 10-20/22-38-43-51/53
Flammable.Harmful by inhalation and if swallowed.Irritating to skin.May cause sensitisation by skin contact.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 24-37-41-61
Avoid contact with skin.Wear suitable gloves.In case of fire and/or explosion do not breathe fumes.Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?