• USA Home
  • 8.20198 - 1-Bromo-4-fluorobenzene

EMAIL THIS PAGE TO A FRIEND
8.20198 EMD Millipore

1-Bromo-4-fluorobenzene

1-Bromo-4-fluorobenzene for synthesis. CAS 460-00-4, molar mass 175 g/mol., for synthesis

  • CAS Number 460-00-4

  • Linear Formula C₆H₄BrF

  • Molecular Weight 175 g/mol

  •  EC Number 207-300-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
bp   151 - 163 °C (1013 hPa)
vapor pressure   25 hPa (50 °C)
solubility   0.136 g/l
density   1.60 g/cm3 (20 °C)
flash point   53 °C
mp   -16 °C
LD 50 oral   LD50 Rat 2700 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.598 - 1.602
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2903 99 80
Hazard Statement(s) 
H226: Flammable liquid and vapour.
H332: Harmful if inhaled.
Precautionary Statement(s) 
P210: Keep away from heat.
Storage class 
3 Flammable liquids
WGK 
WGK 1 slightly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Hazard Pictogram(s) 
GHS02  GHS02
Hazard Pictogram(s) 
GHS07  GHS07
Hazard Symbols 
Harmful  Harmful
Declaration (railroad and road) ADR, RID 
UN 1993 , 3, III
Declaration (transport by air) IATA-DGR 
UN 1993 , 3, III
Declaration (transport by sea) IMDG-Code 
UN 1993 , 3, III
Categories of danger 
flammable, harmful
R Phrase 
R 10-20
Flammable.Harmful by inhalation.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?