• USA Home
  • 8.20211 - 1-Bromo-4-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
8.20211 EMD Millipore

1-Bromo-4-nitrobenzene

1-Bromo-4-nitrobenzene for synthesis. CAS 586-78-7, chemical formula 1-(Br)-4-(NO₂)C₆H₄., for synthesis

  • CAS Number 586-78-7

  • Linear Formula C₆H₄BrNO₂

  • Molecular Weight 202 g/mol

  •  EC Number 209-583-8

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Glass bottle (8202110025)
bp   252.6 °C (1013 hPa)
flash point   >110 °C
mp   123 - 126 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 123 °C
  Melting range (upper value): ≤ 126 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2904 99 00
Storage class 
6.1C Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Declaration (railroad and road) ADR, RID 
UN 3459 , 6.1, III
Declaration (transport by air) IATA-DGR 
UN 3459 , 6.1, III
Declaration (transport by sea) IMDG-Code 
UN 3459 , 6.1, III
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?