• USA Home
  • 8.20216 - DL-α-Bromophenylacetic acid

EMAIL THIS PAGE TO A FRIEND
8.20216 EMD Millipore

DL-α-Bromophenylacetic acid

DL-α-Bromophenylacetic acid for synthesis. CAS 4870-65-9, chemical formula C₆H₅CHBrCOOH., for synthesis

  • CAS Number 4870-65-9

  • Linear Formula C₈H₇BrO₂

  • Molecular Weight 215.04 g/mol

  •  EC Number 225-477-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   25 g in Plastic bottle (8202160025)
mp   78 - 82 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (acidimetric): ≥ 98.0 %
  Melting range (lower value): ≥ 78 °C
  Melting range (upper value): ≤ 82 °C
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2916 39 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
4
Liquid organic acids or solutions should be diluted, if necessary, and carefully neutralized with sodium hydrogen carbonate (Cat. No. 106323) or with sodium hydroxide (Cat. No 106462). Before filling into container D, check the pH with pH universal indicator strips (Cat. No. 109535).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?