• USA Home
  • 8.20217 - 4-Bromophenylhydrazinium chloride

EMAIL THIS PAGE TO A FRIEND
8.20217 EMD Millipore

4-Bromophenylhydrazinium chloride

4-Bromophenylhydrazinium chloride for synthesis. CAS 622-88-8, chemical formula 4-(Br)C₆H₄NHNH₂ * HCl., for synthesis

  • CAS Number 622-88-8

  • Linear Formula C₆H₈BrClN₂

  • Molecular Weight 223.50 g/mol

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   10 g in Glass bottle (8202170010)
mp   200 - 205 °C Decomposition
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (argentometric): ≥ 97.0 %
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2928 00 90
Hazard Statement(s) 
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled.
Precautionary Statement(s) 
P280: Wear protective gloves/ protective clothing.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P304 + P340: IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
RTECS 
MV0800000
Storage class 
6.1A Combustible substances, toxic
WGK 
WGK 3 highly water endangering
Disposal 
9
Carcinogenic compounds and flammable compounds labelled "Highly toxic" or "Toxic": container F. Alkyl sulfates are carcinogenic; Take particular care to avoid inhalation and skin contact. To neutralize alkyl sulfates, add dropwise (from a dropping funnel) to concentrated ice-cool Ammonia solution (Cat.No. 105426) with vigorous stirring. Before placing in container D, check the pH with pH Universal indicator strips (Cat.No. 109535).
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Symbols 
Toxic  Toxic
Categories of danger 
toxic
R Phrase 
R 23/24/25
Toxic by inhalation, in contact with skin and if swallowed.
S Phrase 
S 36/37-45
Wear suitable protective clothing and gloves.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?