EMAIL THIS PAGE TO A FRIEND
8.20249 EMD Millipore

2-tert-Butylphenol

2-tert-Butylphenol for synthesis. CAS No. 88-18-6, EC Number 201-807-2., for synthesis

  • CAS Number 88-18-6

  • Linear Formula C₁₀H₁₄O

  • Molecular Weight 150.22 g/mol

  •  EC Number 201-807-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8202490005)
vapor pressure   20.6 hPa (25 °C)
solubility   0.97 g/l soluble
density   0.98 g/cm3 (20 °C)
flash point   84 °C
autoignition temp.   355 °C
LD 50 oral   LD50 Rat > 200 - 2000 mg/kg
LD 50 dermal   LD50 Rat 705 mg/kg
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 0.981 - 0.983
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2907 19 90
Hazard Statement(s) 
H302 + H332: Harmful if swallowed or if inhaled
H311: Toxic in contact with skin.
H314: Causes severe skin burns and eye damage.
H411: Toxic to aquatic life with long lasting effects.
Precautionary Statement(s) 
P273: Avoid release to the environment.
P280: Wear protective gloves/ protective clothing/ eye protection/ face protection.
P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
P302 + P352: IF ON SKIN: Wash with plenty of soap and water.
P305 + P351 + P338: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P308 + P310: IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician.
Storage class 
6.1 C Combustible, acute toxicity cat. 3 / toxic or substances with chronic effects
WGK 
WGK 2 water endangering
Disposal 
1
Strongly contaminated halogen-free organic solvents: container A.
Hazard Pictogram(s) 
GHS05  GHS05
Hazard Pictogram(s) 
GHS06  GHS06
Hazard Pictogram(s) 
GHS09  GHS09
Hazard Symbols 
Corrosive  Corrosive
Hazard Symbols 
Dangerous for the environment  Dangerous for the environment
Declaration (railroad and road) ADR, RID 
UN 3145 , 8, III
Declaration (transport by air) IATA-DGR 
UN 3145 , 8, III
Declaration (transport by sea) IMDG-Code 
UN 3145 , 8, III
Categories of danger 
harmful, corrosive, dangerous for the environment
R Phrase 
R 20/21/22-34-51/53
Harmful by inhalation, in contact with skin and if swallowed.Causes burns.Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
S Phrase 
S 26-36/37/39-45-61
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.Wear suitable protective clothing, gloves and eye/face protection.In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).Avoid release to the environment. Refer to special instructions/ Safety data sheets.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?