EMAIL THIS PAGE TO A FRIEND
8.20262 EMD Millipore

Quinoxaline

Quinoxaline for synthesis. CAS 91-19-0, molar mass 130.14 g/mol., for synthesis

Synonym: Benzopyrazine, 1,4-Benzodiazine

  • CAS Number 91-19-0

  • Linear Formula C₈H₆N₂

  • Molecular Weight 130.14 g/mol

  •  EC Number 202-047-4

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 g in Glass bottle (8202620100)
solubility   660 g/l soluble
density   1.133 g/cm3 (45 °C)
flash point   98 °C
mp   29 - 32 °C (External MSDS)
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Melting range (lower value): ≥ 30 °C
  Melting range (upper value): ≤ 33 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2933 99 80
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 2 water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?