EMAIL THIS PAGE TO A FRIEND
8.20274 EMD Millipore

4-Chloroanisole

4-Chloroanisole for synthesis. CAS 623-12-1, chemical formula 4-(Cl)C₆H₄OCH₃., for synthesis

  • CAS Number 623-12-1

  • Linear Formula C₇H₇ClO

  • Molecular Weight 142.58 g/mol

  •  EC Number 210-772-2

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   100 ml in Glass bottle (8202740100)
density   1.17 g/cm3 (20 °C)
flash point   75 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 98.0 % (a/a)
  Density (d 20 °C/ 4 °C): 1.171 - 1.173
  Identity (IR): passes test

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2909 30 90
Storage class 
10 - 13 Other liquids and solids
WGK 
WGK 3 highly water endangering
Disposal 
2
Organic halogenated solvents and solutions of organic halogen-containing substances: For strongly contaminated or mixed halogenated solvents container B: Caution: Do not use aluminium containers; in addition, in the case of water-containing chlorinated waste materials do not use stainless-steel containers (danger of leaks caused by corrosion).
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?