• USA Home
  • 8.20279 - 2-Chlorobenzothiazole

EMAIL THIS PAGE TO A FRIEND
8.20279 EMD Millipore

2-Chlorobenzothiazole

2-Chlorobenzothiazole for synthesis. CAS 615-20-3, molar mass 169.62 g/mol., for synthesis

  • CAS Number 615-20-3

  • Linear Formula C₇H₄ClNS

  • Molecular Weight 169.62 g/mol

  •  EC Number 210-415-0

Purchase

Properties

Related Categories Biochemicals and Reagents More...
packaging   5 ml in Glass bottle (8202790005)
bp   140 - 142 °C (40 hPa)
density   1.37 g/cm3 (20 °C)
flash point   >112 °C
mp   21 - 24 °C
shipped in   ambient
storage temp.   Store below +30°C.
specifications   Assay (GC, area%): ≥ 99.0 % (a/a)
  Melting range (lower value): ≥ 21 °C
  Melting range (upper value): ≤ 24 °C
  Identity (IR): passes test
  Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Price and Availability


Ask us for bulk and customization needs

My Chemicals
Safety & Documentation

Safety Information

HS Code 
2934 20 80
Storage class 
10 Combustible liquids not in Storage Class 3
WGK 
WGK 3 highly water endangering
Disposal 
3
Relatively unreactive organic reagents should be collected in container A. If halogenated, they should be collected in container B. For solid residues use container C.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?